Skip to Content
Merck
All Photos(1)

Key Documents

14-711-M

Sigma-Aldrich

Casein Kinase 1γ1 Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32
Product 14-711-M is not currently sold in your country. Contact Technical Service

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

description

protein target CK1γ1

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... CSNK1G1(53944)

General description

FUNCTION: SwissProt: Q9HCP0 # Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling (By similarity).
SIZE: 422 amino acids; 48511 Da
SUBUNIT: Monomer (By similarity).
SUBCELLULAR LOCATION: Cytoplasm.
PTM: Autophosphorylated (By similarity).
SIMILARITY: SwissProt: Q9HCP0 ## Belongs to the protein kinase superfamily. CK1 Ser/Thr protein kinase family. Casein kinase I subfamily. & Contains 1 protein kinase domain.

Packaging

Also available in 10 μg size (cat#14-711) and in bulk (cat#14-711-K).

Quality

Routinely evaluated by phosphorylation of 200μM KRRRALS(p)VASLPGL
routinely evaluated by phosphorylation of 200μM KRRRALS(p)VASLPGL

Physical properties

41.9kDa

Storage and Stability

6 months at -70°C from date of shipment

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service