Skip to Content
Merck
All Photos(1)

Key Documents

1.46423

Millipore

ReadyTube® Violet Red Bile Lactose Agar

For the detection of coliforms in food samples in accordance to ISO 4832, pkg of (200 mL in 250 mL bottle with blue screw cap and 3 loci (6 bottles per box))

Synonym(s):

VRBL Agar

Sign Into View Organizational & Contract Pricing

Select a Size

6 PKG
$81.60

$81.60


Estimated to ship on01 June 2025


Request more information

Select a Size

Change View
6 PKG
$81.60

About This Item

UNSPSC Code:
41106225
NACRES:
NA.74

$81.60


Estimated to ship on01 June 2025


Request more information

Agency

ISO 4832

Quality Level

feature

ready-to-use

manufacturer/tradename

ReadyTube®

packaging

pkg of (200 mL in 250 mL bottle with blue screw cap and 3 loci (6 bottles per box))

technique(s)

cell counting: suitable

pH

7.4 ( in H2O)

application(s)

food and beverages
industrial qc
microbiology

storage temp.

2-25°C

suitability

coliforms

General description

ReadyTube® is ready to use media to provide the highest level of quality and testing confidence for improving accuracy and significantly reducing the risk of false results. ReadyTube® Violet Red Bile Lactose Agar is used for the detection and colony counting of coliform bacteria. The enzymatic digest of animal tissues and yeast extract provide nitrogenous compounds and other nutrients essential for bacterial metabolism. This media is selective due to the presence of the bile salts and crystal violet. Crystal violet inhibits Gram-positive organisms, especially Staphylococci. Neutral red indicator helps to detect lactose fermentation. Lactose fermenting strains grow red or pink and are surrounded by a zone of acid precipitated bile. Sodium chloride maintains the osmotic equilibrium in the medium. The red color is due to absorption of neutral red and a subsequent color change of the dye when the pH of the medium falls below 6.8.

Application

ReadyTube® Violet Red Bile Lactose Agar is recommended for the detection and enumeration of coliform organisms in water, milk, dairy, and other food products.

Legal Information

READYTUBE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service