Skip to Content
Merck
All Photos(1)

Key Documents

1.46365

Millipore

Plate Count Agar

Determination of the total viable count of aerobic bacteria from milk, dairy products, water, and other sample materials.

Synonym(s):

Tryptone Glucose Yeast Agar, Casein-Peptone Dextrose Yeast Agar, Standard Methods Agar, PCA

Sign Into View Organizational & Contract Pricing

Select a Size

6 PKG
$70.90

$70.90


Estimated to ship on31 May 2025


Request more information

Select a Size

Change View
6 PKG
$70.90

About This Item

UNSPSC Code:
41106225
NACRES:
NA.74

$70.90


Estimated to ship on31 May 2025


Request more information

Quality Level

form

(Solid Medium)

feature

ready-to-use

packaging

pkg of (200 ml in 250 ml bottle with blue screw cap and 3 loci (6 bottles per box))

pH

7.0 ( in H2O)

application(s)

food and beverages
industrial qc
microbiology

storage temp.

2-25°C

suitability

nonselective for

General description

Plate Count Agar medium is suitable for the enumeration of the total viable count. The pH is in the optimum range of 6.8-7.2 for the growth of bacteria. The media is relatively rich in its nutrients and does not contain any inhibitors or indicators. The enzymatic digest of casein serves as a nitrogen source with high levels of free amino acids followed by yeast extract, which is a rich source of vitamins. Glucose serves as the carbon source for the efficient growth of bacteria.

Application

Plate Count Agar medium is used for the enumeration of the total viable microbial count on a wide variety of food, water and beverage products. e.g. dairy products, bottled water, fruit juices etc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service