Skip to Content
Merck
All Photos(2)

Key Documents

1.46189

Millipore

Baird-Parker Agar

ReadyPlate, ISO 6888, FDA BAM, APHA, plate diam. 55 mm, box of 20 plates, for Staphylococcus spp.

Synonym(s):

ReadyPlate CT Baird-Parker Agar ISO 6888, Staphylococcus Selective Agar

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
$94.70

$94.70


Estimated to ship on01 June 2025


Request more information

Select a Size

Change View
20 EA
$94.70

About This Item

UNSPSC Code:
41106231
NACRES:
NA.74

$94.70


Estimated to ship on01 June 2025


Request more information

Agency

APHA
ISO 6888

Quality Level

reg. compliance

FDA BAM

product line

ReadyPlate

form

plate

packaging

box of 20 plates

manufacturer/tradename

ReadyPlate

plate diam.

55 mm

pH

6.8 ( in H2O)

application(s)

food and beverages
surface monitoring

storage temp.

15-25°C

suitability

Staphylococcus spp.
bacteria

General description

ReadyPlate CT Baird-Parker Agar contact plates provide a ready-to-use method for environmental monitoring of surfaces in critical clean rooms and isolators.
The Baird-Parker medium attains its selectivity with potassium chloride, lithium chloride, and glycine. Sodium pyruvate is essential to both recovery of damaged Staphylococcus aureus cells and their subsequent growth. Agar is the solidifying agent.
With added egg yolk tellurite emulsion, coagulase-positive staphylococci form black or grey colonies due to tellurite reduction with or without egg yolk reaction. Egg-yolk reaction is shown by characteristic zones and rings formed as a result of lipolysis and/or proteolysis.
According to EN ISO 6888-1, the addition of sulfamethazine is advised to suppress the growth and swarming of Proteus spp. if these are suspected in the test sample.

Application

ReadyPlate CT Baird-Parker Agar contact plates are suitable for the isolation, presumptive identification, and enumeration of Staphylococcus aureus from sampled surfaces as well as other materials like food and animal feed.

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Contact Plates and Swabs for Personnel and Surface Monitoring in Cleanrooms and Isolators

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service