Skip to Content
Merck
All Photos(1)

Key Documents

14-601-M

Sigma-Aldrich

ROKβ/ROCK-I Protein, active, 10 µg

Active, N-terminal His6-tagged, recombinant, human ROCK-I amino acids 17-535, autoactivated by incubating with Mg/ATP & redialysed to remove excess ATP. For use in Kinase Assays.

Synonym(s):

Rho-associated protein kinase, p160ROCK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41
Product 14-601-M is not currently sold in your country. Contact Technical Service

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

form

liquid

shelf life

6 mo.

mol wt

Mw 61.4 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

storage temp.

−70°C

Gene Information

human ... ROCK1(6093)

General description

Research area: Apoptosis

N-terminal His6-tagged, recombinant, human ROCK-I amino acids 17-535; autoactivated by incubating with Mg/ATP and redialysed to remove excess ATP

Product Source: expressed by baculovirus in Sf21 insect cells

Application

Research Category
Inflammation & Immunology

Biochem/physiol Actions

G protein, RhoA/Rho kinase 1 (ROCK1) [1] is a serine/threonine kinase. It plays a vital role in the regulation of cell growth, and invasive and metastatic progression of various types of human cancers. ROCK1 is involved in the regulation of apoptosis or programmed cell death in various cell types.[2] Overexpression of ROCK1 has been observed in various tumors, including glioblastoma, melanoma, osteosarcoma, and hepatocellular carcinoma. This protein is known to enhance actin-myosin contractility by phosphorylating regulatory myosin light chains (MLCs). It also plays a role in cell motility through the modulation of integrin β1-activated focal adhesion kinase (FAK) signaling. [3]

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-601M when ordering the 250 µg size.

Quality

routinely evaluated by phosphorylation of Long S6 substrate peptide (Catalog #12-420)

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service