Skip to Content
Merck
All Photos(1)

Key Documents

14-583-M

Sigma-Aldrich

FGFR4 Protein, active, 10 µg

Active, N-terminal, His6-tagged, recombinant, human FGFR4 amino acids 442-755, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
eCl@ss:
32160405
NACRES:
NA.32
Product 14-583-M is not currently sold in your country. Contact Technical Service

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 36.4 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

Gene Information

human ... FGFR4(2264)

General description

N-terminal, His6-tagged, recombinant, human FGFR4 amino acids 442-755

Biochem/physiol Actions

Protein Target: FGFR4

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-583M when ordering the 250 µg size.

Quality

routinely evaluated by phosphorylation of Poly (Glu4-Tyr) (4:1)

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service