14-439-M
MAP Kinase 1/Erk1 Protein, active, 10 µg
Active, recombinant, full-length, human MAP Kinase 1/Erk1 containing an N-terminal GST tag, for use in Kinase Assays.
Synonym(s):
ERT1, Extracellular signal-regulated kinase 2 (ERK-2), MAPK 1, Mitogen-activated protein kinase 1, p44 MAPK
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in E. coli
form
liquid
mol wt
Mw 69.9 kDa
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (kinase)
color
colorless
solubility
water: soluble
NCBI accession no.
UniProt accession no.
Gene Information
human ... MAPK3(5595)
General description
Research area: Cell Signaling
Recombinant, full-length, human MAP Kinase 1/Erk1 containing an N-terminal GST tag
Recombinant, full-length, human MAP Kinase 1/Erk1 containing an N-terminal GST tag
Biochem/physiol Actions
Extracellular-signal-regulated kinase 1 (ERK1)/ Extracellular-signal-regulated kinase 2 (ERK2), as part of the Ras-Raf-MEK-ERK signal transduction cascade, are involved in regulating a wide range of cellular processes, including cell adhesion, cell cycle progression, cell migration, cell survival, differentiation, metabolism, proliferation, and transcriptional control. Additionally, ERK1 signaling contributes to the regulation of cyclin D1 expression during the cell cycle. MAP kinases play a crucial role in inflammation and immune responses by regulating the production of pro- and anti-inflammatory cytokines, as well as prostaglandins, in response to lipopolysaccharide (LPS) stimulation.
Packaging
Also available in 250μg size as catalog # 14-439M - Call for pricing and availability.
Quality
routinely evaluated by phosphorylation of MBP
Physical form
Purified by glutathione-agarose. Activated with MEK1 and repurified on Ni2+/NTA agarose
Other Notes
For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of biological chemistry, 276(24), 20913-20923 (2001-03-30)
Ultraviolet light A (UVA) plays an important role in the etiology of human skin cancer, and UVA-induced signal transduction has a critical role in UVA-induced skin carcinogenesis. The upstream signaling pathways leading to p70(S6K) phosphorylation and activation are not well
The Journal of biological chemistry, 279(18), 18239-18246 (2004-02-07)
Vav1 is a signaling protein required for both positive and negative selection of CD4(+)CD8(+) double positive thymocytes. Activation of the ERK MAPK pathway is also required for positive selection. Previous work has shown that Vav1 transduces T cell receptor (TCR)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service