Skip to Content
Merck
All Photos(1)

Documents

1.44207

Millipore

Serial Cable

For RCS® High Flow Touch, connection via RS232 interface

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26121630
NACRES:
NA.74

material

plastic cable(s)

Agency

non-compendial

feature

electronic
portable

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100 VF®

storage temp.

room temp

General description

Serial cable is an additional accessory for use with RCS® High Flow Touch microbial air sampler. The RCS® High Flow Touch is a portable air sampler for fast and easy monitoring of ambient air and compressed gases in controlled environments. This microbial air sampler is based on impaction by centrifugal force and can also be used with a compressed gas adapter for additional monitoring of compressed gases. The cable is used for connecting RCS® Instrument to a PC via the integrated, serial RS232 interface

Application

Serial cable is an additional accessory with the RCS® High Flow air sampler for connecting RCS® Instrument to a PC via the integrated, serial RS232 interface. These air samplers find application in microbial monitoring of ambient air in the pharmaceutical, medical, and food industries.

Legal Information

MAS-100 VF is a registered trademark of MBV AG
RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service