Skip to Content
Merck
All Photos(5)

Key Documents

1.32359

Millipore

MC-Media Pad®

for rapid aerobic count

Synonym(s):

Rapid Aerobic Count test, RAC test

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$299.00

$299.00


Available to ship on31 March 2025Details


Request more information

Select a Size

Change View
1 EACH
$299.00

About This Item

UNSPSC Code:
41171504
NACRES:
NA.75

$299.00


Available to ship on31 March 2025Details


Request more information

Agency

AOAC® OMA
AOAC® PTM
MicroVal

product line

MC-Media Pad®

feature

ready-to-use

storage condition

(Store upright)

technique(s)

cell counting: suitable (total microbial count)

pH

7.2 ( in H2O)

application(s)

food and beverages
personnel monitoring
surface monitoring
water monitoring

shipped in

ambient

storage temp.

2-25°C

suitability

aerobic bacteria

General description

The MC MEDIA PAD® offers a convenient solution for rapidly and routinely testing microbial contamination in raw and in-process food and beverage materials, as well as finished products. The MC MEDIA PAD® is a pre-sterilized, ready-to-use dry culture device, simplifying testing and minimizing the quantity of waste. MC MEDIA PAD® is composed of a unique adhesive sheet, a test pad coated with medium and water absorption polymer, and a transparent cover film, to automatically re-constitute the media. It is intended to determine the total aerobic count using a special medium composition and unique redox indicator dyes for not only standard but also rapid enumeration. The MC MEDIA PAD® is coated with growth medium and chromogenic substrate for total aerobic enumeration. Once the liquid sample is inoculated onto the test pad, the sample diffuses through the whole pad by capillary action.

Application

The MC MEDIA PAD® enables rapid detection of total aerobic microbial contamination and therefore is very useful in checking for microbial contamination in raw and in-process food and beverage samples. While improving the workflow, it also ensures accurate and reliable results.

Features and Benefits

  • Save space in fridge and incubator
  • Go green & gain: reduce environmental impact
  • Improve the inventory management with a shelf-life of up to 24 months
  • Comply with regulations
  • No spreading device required
  • Simplified workflow

Analysis Note

If aerobic bacteria are present, they grow as red coloured colonies on test pad.

Legal Information

AOAC is a registered trademark of AOAC International
MC MEDIA PAD is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This article will focus on results obtained for MC-Media Pad RAC, MC-Media Pad Coliform, MC-Media Pad E. coli & Coliform & MC-Media Pad Yeast & Mold.

Updates on ISO standards in food microbiology by expert Barbara Gerten from Merck

Protocols

Comparison of MC-Media Pad® with different convenience solution products for indicator organism testing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service