Skip to Content
Merck
All Photos(1)

Key Documents

1.30196

Millipore

HY-LiTE® Jet A1 Fuel Test Kit

Synonym(s):

Jet A1 Fuel biomass test, Rapid Jet A1 fuel test

Sign Into View Organizational & Contract Pricing

Select a Size

20 TEST
$1,220.00

$1,220.00


Available to ship on01 April 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
20 TEST
$1,220.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$1,220.00


Available to ship on01 April 2025Details


Request a Bulk OrderRequest more information

description

HY-LiTE® Jet A1 Fuel Test Kit is suitable for rapid detection of microbiological contamination in fuel tanks

product line

HY-LiTE®

shelf life

15 mo.

packaging

pkg of 20 tests (20 pens, 20 large pipettes, 20 small pipettes)

manufacturer/tradename

HY-LiTE®

storage condition

dry at room temperature

application(s)

jet fuel

compatibility

for use with HY-LITE®

detection method

bioluminescent

storage temp.

2-8°C

General description

The HY-LiTE® Jet A1 Fuel Test Kit contains ready-to-use pens for detection of total biological contamination of Jet A1 fuel used in conjunction with the HY-LiTE® 2luminometer. Fuel contains small amounts of water and bears the risk of microbial contamination. Adenosine triphosphate (ATP) is detected by reaction with a luciferin/luciferase reagent in a buffered solution. The test kit is designed to indicate the presence of metabolically active biological material such as bacteria, yeast, and molds, including sulphate-reducing bacteria and other anaerobic microorganisms, which may not be detected by traditional growth-based methods.MEASUREMENT LIMITS:
IATA (International Air Transport Association) “Guidance Material on Microbiological Contamination in Aircraft Fuel Tanks” 5th Edition:
  • Negligible: < 1000 RLU/liter fuel
  • Moderate: 1000 – 5000 RLU/liter fuel
  • Heavy: > 5000 RLU/liter fuel
The limits suggested above are based on the HY-LiTE® 2 luminometer’s measurement scale. They apply only to Aircraft Wing Tank samples and are based on comparison with other routinely used methods. These limits are a suggestion only and may need to be adapted for other types of samples and according to individual circumstances, frequency of testing, history of contamination etc.
This fuel test kit contains patented capture solution for sampling from Jet A1 fuel samples.

Application

The HY-LiTE® Jet A1 Fuel Test Kit is suitable for rapid detection of microbiological contamination in fuel tanks and distribution systems.

Features and Benefits

  • Results obtained within minutes
  • De-skilled method
  • Field test - no need for laboratory facilities
  • Quantitative, Objective results
  • Numerical read-out. Easy interpretation
  • Same protocol for all jet fuel samples (with / without water)
  • Same action limits for all samples (with / without water)
  • Detects biological activity directly in the sample and does not require the growth of microorganisms in laboratory media
  • Flexible sample volume. 1 liter recommended but lower volumes can be tested. For comparison with guidelines, results can be volume adjusted: RLU/liter = RLU x (1000 ml/ml sample volume)Recommended in IATA Guidance Material on Microbiological Contamination in Aircraft Fuel Tanks 5th Ed.

Suitability

For examination of biomass contamination of Jet A1 and other fuel samples

Analysis Note

Packaging: Passes test
Appearance: Passes test
Performance test: Passes test

Legal Information

HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service