Skip to Content
Merck
All Photos(1)

Key Documents

1.25156

Millipore

Eshmuno® CPX Resin

MiniChrom Column Eshmuno® CPX, volume 1 mL, prepacked column

Synonym(s):

MiniChrom Column Eshmuno®CPX, 1ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
Pricing and availability is not currently available. Contact Technical Service

ligand

(Sulfoisobutyl)

Quality Level

material

polypropylene housing

product line

Eshmuno®

form

prepacked column

parameter

≤8 bar pressure
500 cm/hr max. flow rate

column I.D. × L

8 mm × 20 mm

volume

1 mL

matrix active group

Polyvinyl Ether

mean particle size

50 μm

capacity

~120 mL, resin binding capacity

application(s)

mAb
recombinant protein
vaccine development

separation technique

cation exchange

storage temp.

no temp limit

Looking for similar products? Visit Product Comparison Guide

General description

8 X 20mm

Features and Benefits

Eshmuno® CPX chromatography resin enables:

  • Intermediate purification or polishing of monoclonal antibodies
  • High aggregate removal efficiency combined with high dynamic binding capacity
  • High loading capacity combined with high purity at lower elution volumes.
  • Wide pH and conductivity operating window
  • High flow rate versus pressure-flow behavior
  • Excellent removal of impurities
  • Robust and safe packing procedures
  • Strong chemical stability
  • Compatibility with 1 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl and storage solutions 0.1 M NaOH

Packaging

Plastic bag(s)

Legal Information

Eshmuno is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn more on mAb downstream processing, more specifically polishing chromatography and the relevant associated products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service