Skip to Content
Merck
All Photos(1)

Key Documents

12-493

Sigma-Aldrich

Abltide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$477.00

$477.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$477.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

$477.00


Estimated to ship on26 May 2025


Request a Bulk Order

manufacturer/tradename

Upstate®

Quality Level

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Biochem/physiol Actions

Protein Target: Abl
Target Sub-Family: TK

Quality

Routinely evaluated for use as a substrate for in vitro Abl-family kinase assays.

Physical form

Lyophilized powder

Storage and Stability

Lyophilized: 2 years at 4°C. Rehydrated: 6 months at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service