Skip to Content
Merck
All Photos(1)

Key Documents

1.24505

Sigma-Aldrich

Osmium(VIII) oxide

for microscopy

Synonym(s):

Osmium(VIII) oxide, Osmic acid, Osmiumtetraoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
O4Os
CAS Number:
Molecular Weight:
254.23
MDL number:
UNSPSC Code:
12352303
EC Index Number:
244-058-7

vapor pressure

10 hPa ( 20 °C)

Quality Level

form

solid

potency

15 mg/kg LD50, oral (Rat)

color

yellow

pH

6 (20 °C, 25 g/L in H2O)

bp

130 °C/1013 hPa

mp

40 °C

solubility

65 g/L

density

4.91 g/cm3 at 20 °C

bulk density

1500 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

InChI

1S/4O.Os

InChI key

VUVGYHUDAICLFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Osmium tetroxide for microscopy can be used to visualize unsaturated lipids. A part of the lipids is immediately blackened by the influence of osmic acid solution. The immediate black coloration results from the -C=C- double bond of the unsaturated fatty acid. The fatty acids that are made brownish at first can be blackened by treatment with 60-70% alcohol. For electron microscopy or semithin sectioning the glutardialdehyde-fixed specimens are post-fixed with osmium(VIII) oxide. Lipid inclusions appear black on a yellow to brown background. It is an IVD / CE registered product. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Linkage

Replaces: OX0260-5; OX0260-3

Analysis Note

Identity (Raman-spectrum): conforms

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoji Horigome et al.
Autophagy, 16(3), 501-511 (2019-06-18)
Chondrogenesis is accompanied by not only cellular renovation, but also metabolic stress. Therefore, macroautophagy/autophagy is postulated to be involved in this process. Previous reports have shown that suppression of autophagy during chondrogenesis causes mild growth retardation. However, the role of
Charles C H Cohen et al.
Cell, 180(2), 311-322 (2019-12-31)
The propagation of electrical impulses along axons is highly accelerated by the myelin sheath and produces saltating or "jumping" action potentials across internodes, from one node of Ranvier to the next. The underlying electrical circuit, as well as the existence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service