Skip to Content
Merck
All Photos(1)

Key Documents

12-404

Sigma-Aldrich

Histone H3 Peptide

biotin conjugate, residues 21-44

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$551.00

$551.00


Estimated to ship on25 May 2025



Select a Size

Change View
100 μG
$551.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

$551.00


Estimated to ship on25 May 2025


Product Name

Histone H3 Peptide, biotin conjugate, residues 21-44, This gene contains introns & its mRNA is polyadenylated, unlike most histone genes. The protein encoded is a replication-independent member of the histone H3 family.

Quality Level

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... H3F3B(3021)

Application

This gene contains introns & its mRNA is polyadenylated, unlike most histone genes. The protein encoded is a replication-independent member of the histone H3 family.

Quality

Routinely evaluated by as a substrate for histone-modifying enzymes.

Physical form

Lyophilized

Storage and Stability

Lyophilized: 2 years at 4°C. Rehydrated: 6 months at -20°C.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DNA replication cycle in parthenogenetically developing eggs of the starfish Asterina pectinifera.
A Nomura,S Nemoto
Development, Growth & Differentiation null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service