Skip to Content
Merck
All Photos(1)

Key Documents

12-334

Sigma-Aldrich

Rabbit Anti-Chicken IgY, bridging Antibody

Upstate®, from rabbit

Synonym(s):

Chicken IgG

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$263.00

$263.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
250 μG
$263.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.47

$263.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

chicken

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

shipped in

dry ice

target post-translational modification

unmodified

Application

Rabbit anti-Chicken IgY Antibody, bridging is an antibody against Chicken IgY for use in IP & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Quality

Routinely evaluated in a western blot.

Physical form

0.01M sodium phosphate, pH 7.4, 0.14M sodium chloride

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service