Skip to Content
Merck
All Photos(1)

Key Documents

119139

Sigma-Aldrich

Adenosine A3 Receptor Agonist, 2-Cl-IB-MECA

A highly selective agonist of adenosine A3 receptor (Ki = 330 pM)

Synonym(s):

Adenosine A3 Receptor Agonist, 2-Cl-IB-MECA

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$461.00

$461.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
2 MG
$461.00

About This Item

Empirical Formula (Hill Notation):
C18H18ClIN6O4
CAS Number:
Molecular Weight:
544.73
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$461.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Ic1cc(ccc1)CNc2nc(nc3[n](cnc32)[C@@H]4O[C@@H]([C@H]([C@H]4O)O)C(=O)NC)Cl

InChI

1S/C18H18ClIN6O4/c1-21-16(29)13-11(27)12(28)17(30-13)26-7-23-10-14(24-18(19)25-15(10)26)22-6-8-3-2-4-9(20)5-8/h2-5,7,11-13,17,27-28H,6H2,1H3,(H,21,29)(H,22,24,25)/t11-,12+,13-,17+/m0/s1

InChI key

IPSYPUKKXMNCNQ-PFHKOEEOSA-N

General description

A highly selective agonist of adenosine A3 receptor (Ki = 330 pM)
An adenosine analog that acts as a highly selective agonist of adenosine A3 receptor (Ki = 330 pM, 820 nM, 470 nM for A3, A1 and A2A, respectively). Shown to reduce ischemia reperfusion injury in mice and limit infarct size in isolated rat hearts. Shown to reversibly increase field excitatory postsynaptic potentials in hippocampal slices and exhibits excitatory effects on both evoked and spontaneous epileptiform discharges.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kim, H. O., et al. 1994. J Med Chem.37, 3614.
Laudadio, M. A., et al. 2004. Epilepsy Res.59, 83.
Ge, Z.D., et al. 2006. J. Pharm. Exp. Ther.319, 1200.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service