Skip to Content
Merck
All Photos(2)

Key Documents

1.18324

Supelco

Turbidimeter

turbidity 1100 NTU

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$2,750.00

$2,750.00


Available to ship on31 March 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
1 UNIT
$2,750.00

About This Item

UNSPSC Code:
47101516
NACRES:
SE.11

$2,750.00


Available to ship on31 March 2025Details


Request a Bulk OrderRequest more information

technique(s)

photometry: suitable

turbidity

1100 NTU

compatibility

for use with Turbiquant 1100 IR

storage temp.

2-30°C

General description

The Turbiquant 1100 IR is particularly suited for on-the-spot measurements as it is battery-operated. The robust carry-case and the waterproof casing offer additional protection. The turbidimeter contains an IR-LED (infrared) light source with a wavelength of 860 nm for standard-compliant measurements according to EN ISO 7027. Within seconds, the nephelometric turbidity of a sample (NTU/FNU) can be measured. The instrument can be calibrated using the respective Turbidiquant Calibration Standard Set.
portable, incl. batteries, case, 2 empty cells, manual, handy hints 1100 IR 0.01 - 1100 NTU Turbiquant

Legal Information

TURBIQUANT is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

  1. what is the maximum lower and upper limit i terms of NTU of this instrument? I want to measures turbidity of nanosheets dispersions. is it possible to measure accurately?

    1 answer
    1. This turbidimeter has a measuring range of 0.01 to 1100 NTU and can be used to measure the turbidity of nanosheet dispersions accurately.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service