Skip to Content
Merck
All Photos(1)

Key Documents

1.17283

Supelco

ICP Multi element standard solution XXV

for MS (29 elements in diluted nitric acid), Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Product 1.17283 is not currently sold in your country. Contact Technical Service

grade

reference material

Quality Level

product line

Certipur®

technique(s)

ICP: suitable

application(s)

agriculture
environmental
food and beverages

format

multi-component solution

storage temp.

15-25°C

Packaging

100ml in HD-PE bottle

Analysis Note

Al (aluminium): 10 mg/l
As (arsenic): 10 mg/l
Ba (barium): 10 mg/l
Be (beryllium): 10 mg/l
Bi (bismuth): 10 mg/l
Cd (cadmium): 10 mg/l
Ca (calcium): 10 mg/l
Cs (cesium): 10 mg/l
Cr (chromium): 10 mg/l
Co (cobalt): 10 mg/l
Cu (copper): 10 mg/l
Ga (gallium): 10 mg/l
In (indium): 10 mg/l
Fe (iron): 10 mg/l
Pb (lead): 10 mg/l
Li (lithium): 10 mg/l
Mg (magnesium): 10 mg/l
Mn (manganese): 10 mg/l
Ni (nickel): 10 mg/l
K (potassium): 10 mg/l
Rb (rubidium): 10 mg/l
Se (selenium): 10 mg/l
Ag (silver): 10 mg/l
Na (sodium): 10 mg/l
Sr (strontium): 10 mg/l
Tl (thallium): 10 mg/l
U (uranium): 10 mg/l
V (vanadium): 10 mg/l
Zn (zinc): 10 mg/l

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service