Skip to Content
Merck
All Photos(1)

Key Documents

116833

Millipore

Adenosine 3′,5′-cyclic Monophosphate, 8-(4-Chlorophenylthio)-2′-O-Methyl-, Sodium Salt

A potent, cell-permeable, and specific activator of the exchange protein activated by cyclic AMP (EPAC).

Synonym(s):

Adenosine 3′,5′-cyclic Monophosphate, 8-(4-Chlorophenylthio)-2′-O-Methyl-, Sodium Salt, 8-CPT-2ʹ-O-Me-cAMP, Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16ClN5O6PS · Na
Molecular Weight:
507.82
UNSPSC Code:
12352208
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent, cell-permeable, and specific activator of the exchange protein activated by cyclic AMP (EPAC) (also called cAMP-guanine nucleotide-exchange factors or cAMP-GEF) in vitro and in vivo. Does not have any effect on protein kinase A (PKA). A useful tool to discriminate between EPAC and PKA. 5 µmol = 2.54 mg. λmax = 282 nm.
A potent, cell-permeable, and specific activator of the exchange protein activated by cyclic AMP (EPAC) [also called cAMP-guanine nucleotide-exchange factors (cAMP-GEF)] in vitro and in vivo. Does not have any effect on protein kinase A (PKA). A useful tool to discriminate between EPAC and PKA.
Note: 5 µmol = 2.54 mg.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kang, G., et al. 2003. J. Biol. Chem.278, 8279.
Enserink, J.M., et al. 2002. Nat. Cell Biol., 4, 901.

Legal Information

Sold under license of European Patent Application 02/077,219.0.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service