Skip to Content
Merck
All Photos(1)

Key Documents

1.16770

Supelco

TISAB-III solution

Total ionic strength ajustment buffer, for fluoride determination

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$338.00

$338.00


Estimated to ship on08 April 2025


Request more information

Select a Size

Change View
500 ML
$338.00

About This Item

UNSPSC Code:
12352401
NACRES:
NA.21

$338.00


Estimated to ship on08 April 2025


Request more information

Product Name

TISAB-III solution, for fluoride determination

Quality Level

form

liquid

technique(s)

titration: suitable

pH

5.5 (20 °C in H2O)

density

1.093 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Estimation of fluoride uptake in soil and staple food crops produced in highly fluoridated and non-fluoridated regions of Raichur District, Karnataka: This study examined fluoride concentrations in soils and crops from different regions, using methodologies including TISAB-III for accurate fluoride analysis, demonstrating its role in environmental and agricultural research (Havale et al., 2022).

  • Fluoride release from two types of fluoride-containing orthodontic adhesives: Conventional versus resin-modified glass ionomer cements-An in vitro study: This research highlights the use of TISAB-III in dentistry for studying fluoride release from orthodontic adhesives, underscoring its utility in dental material testing (Dziuk et al., 2021).

  • Fluoride and calcium concentrations in the biofilm fluid after use of fluoridated dentifrices supplemented with polyphosphate salts: The application of TISAB-III facilitated the measurement of fluoride and calcium in biofilm fluid in this dental study, which could help in developing better oral care products (Nagata et al., 2017).

  • The determination of fluoride in water in the presence of organic acids and iron using a fluoride-selective electrode: This early study shows the effectiveness of TISAB-III in complex water samples containing organic acids and iron, proving its essential role in environmental chemistry (Nichoison & Pasquier, 1993).

Analysis Note

Concentration (NH₄Cl): 128.0 - 132.0 g/l
Concentration (Titriplex IV): 18.0 - 20.0 g/l
pH-value (20 °C): 5.0 - 5.5
Concentration (fluoride): ≤ 1 µg/ml

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service