Skip to Content
Merck
All Photos(1)

Key Documents

1.16761

Millipore

Thioglycolate medium G

for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74
Pricing and availability is not currently available. Contact Technical Service

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

6.9-7.3 (25 °C, 29 g/L in H2O, after autoclaving)

solubility

29 g/L

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

anaerobic bacteria

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish
Colour (Oxidation area): pink to violet
pH-value (25 °C): 6.9 - 7.3
Growth (Bacillus subtilis ATCC 6633 (WDCM 00003)): +
Growth (Staphylococcus aureus ATCC 6538 (WDCM 00032)): +
Growth (Clostridium sporogenes ATCC 19404 (WDCM 00008)): +
Growth (Clostridium sporogenes ATCC 11437): +
Growth (Bacteroides vulgatus ATCC 8482): +
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): +
Growth (Kocuria rhizophila ATCC 9341): +
Growth (Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)): +
Incubation: up to 3 days; 30 °C to 35 °C; aerobic; Bacteroides up to 5 days.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service