Skip to Content
Merck
All Photos(1)

Key Documents

1.16205

Supelco

Diethanolamine

for analysis EMSURE®

Synonym(s):

Diethanolamine, ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$157.00

$157.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 L
$157.00

About This Item

Linear Formula:
NH(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-868-0
NACRES:
NB.21
grade:
reagent grade
bp:
269-270 °C/1013 hPa
vapor pressure:
<0.01 hPa ( 25 °C)

$157.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

reagent grade

Quality Level

vapor pressure

<0.01 hPa ( 25 °C)

product line

EMSURE®

Assay

≥99.5% (GC)

form

solid

autoignition temp.

365-370 °C

potency

676 mg/kg LD50, oral (Rat)
8328 mg/kg LD50, skin (Rabbit)

expl. lim.

2.1-10.6 % (v/v)

dilution

(for analytical testing)

impurities

≤0.0001% Fe (Iron)
≤0.0001% Heavy metals (as Pb)
≤0.0005% Chloride (Cl)
≤0.002% Sulfate (SO4)
≤0.25% Water (Karl Fischer)
≤0.3% Ethanolamine (GC)
≤0.3% Triethanolamine (GC)

sulfated ash (600 °C)

≤0.005%

pH

11 (20 °C, 53 g/L in H2O)

kinematic viscosity

357.2 cSt(30 °C)

bp

269-270 °C/1013 hPa

mp

28 °C

transition temp

flash point 176 °C

density

1.09 g/cm3 at 30 °C (liquid)

storage temp.

2-30°C

SMILES string

N(CCO)CCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service