1.15092
Extrelut® NT Packing Material Refill pack
packing material for the production of large extraction columns
About This Item
Recommended Products
product name
EXtrelut® NT, packing material for the production of large extraction columns
Quality Level
technique(s)
supported liquid extraction (SLE): suitable
pH
10 (20 °C, 100 g/L in H2O)
mp
>450 °C
solubility
0.0037 g/L
density
2.36 g/cm3 at 20 °C
bulk density
300 kg/m3
storage temp.
15-25°C
InChI
1S/C4H9.Li/c1-3-4-2;/h1,3-4H2,2H3;
InChI key
MZRVEZGGRBJDDB-UHFFFAOYSA-N
Related Categories
General description
Application
- Heart failure biomarkers BNP and NT-proBNP detection using optical labels.: This research utilizes EXtrelut NT for the extraction and analysis of heart failure biomarkers BNP and NT-proBNP. The study highlights the role of EXtrelut NT in enhancing the sensitivity and accuracy of biomarker detection using optical labels, which is critical for the early diagnosis and monitoring of heart failure (Goryacheva et al., 2022).
- Evaluation of the Analytical Performance of BNP and NT-proBNP Assays on the Atellica CI 1900 Analyzer.: This study employs EXtrelut NT to streamline the extraction of BNP and NT-proBNP biomarkers from human plasma and serum samples. The research demonstrates the efficacy of EXtrelut NT in ensuring precise and reproducible results in high-throughput analytical workflows, validating the performance of the Atellica CI 1900 Analyzer in clinical diagnostics (Rhea-McManus et al., 2023).
- NT-proBNP detection with a one-step magnetic lateral flow channel assay.: This study uses EXtrelut NT to facilitate the extraction and purification of NT-proBNP from whole blood samples. The research highlights the role of EXtrelut NT in achieving high sensitivity and accuracy in NT-proBNP detection using a novel magnetic lateral flow assay, which is crucial for point-of-care diagnostics of heart failure (Strohmaier-Nguyen et al., 2024).
- Sample-to-answer lateral flow assay with integrated plasma separation and NT-proBNP detection.: This research employs EXtrelut NT in the development of a sample-to-answer lateral flow assay for NT-proBNP detection. The study emphasizes the pack′s role in ensuring efficient plasma separation and high-sensitivity detection of NT-proBNP in clinical blood samples, highlighting its application in point-of-care testing (Strohmaier-Nguyen et al., 2024).
Analysis Note
Bulk density: 18.0 - 36.0 g/100ml
Alkalinity (Extrelut-20-column): <0.25mmol/E-20-column
purity test
- absorbance 280 nm:
≤ 0.109
- absorbance 250 nm: ≤ 0.157
- absorbance 230 nm: ≤ 0.300
- GC: no to insignificant peaks of contamination
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 1 Inhalation
Target Organs
Lungs
Storage Class Code
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Simplify your liquid-liquid extractions with our emulsion-free supported-liquid extraction (SLE) single step solution.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service