Skip to Content
Merck

1.15079

SAFC

Parteck® SI 200 (Sorbitol)

EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, NF, E 420

Pharma Manufacturing

Synonym(s):

Parteck® Sorbitol, D-Glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352201
E Number:
E420
EC Index Number:
200-061-5
Pricing and availability is not currently available. Contact Technical Service

Agency

BP
JP
NF
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

fine powder

autoignition temp.

420 °C

particle size

(see specification)

pH

6-7 (20 °C, 100 g/L in H2O)

mp

94-96 °C

transition temp

flash point >100 °C

density

1.49 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

2-25°C

SMILES string

OC(C(O)C(O)CO)C(O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2

InChI key

FBPFZTCFMRRESA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With formulation challenges in mind, we have used particle engineering technologies to develop functional excipients specifically for solid dosage forms. The products in our Parteck® portfolio feature unique particle properties tailored for excellent performance in tableting processes, for specific drug delivery technologies, or for solubility enhancement.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Parteck® SI excipient is a directly compressible sorbitol which combines excellent tableting behavior, resulting in very robust and shiny tablets, with a pleasant mouthfeel and taste. The unique and large surface area allows for a fast disintegration and dissolution, even at high tablet hardnesses and supports a good content uniformity of low dose API formulations. Parteck® SI excipient comes in a variety of particle sizes and qualities to meet the needs of a very wide range of applications.

Features and Benefits

  • Excellent direct compression properties
  • High dilution and adsorption capacity
  • Larger and highly structured surface area
  • Low hygroscopicity
  • Low amount of reducing sugars

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
PARTECK is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

This article describes the use of excipients to help address API stability issues.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service