Skip to Content
Merck
All Photos(2)

Key Documents

1.14660

Supelco

Iron Test Kit, colorimetric

0.05-1.0 mg/L (Fe²⁺), MQuant®

Sign Into View Organizational & Contract Pricing

Select a Size

50 TESTS
$91.60

$91.60


Estimated to ship on28 May 2025



Select a Size

Change View
50 TESTS
$91.60

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$91.60


Estimated to ship on28 May 2025


Product Name

Iron Test in freshwater and seawater, colorimetric, 0.05-1.0 mg/L (Fe2+), for use with MQUANT®

specific analyte(s)

iron

Quality Level

measuring range

0.05-1.0 mg/L (Fe2+)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card 0.05 - 0.1 - 0.2 - 0.4 - 0.6 - 0.8 - 1.0 mg/l Fe²⁺ MQuant®
This MQuant® Iron test (formally MColortest®) is suitable for the colorimetric determination using the easy to use color card. The reagents are added to the sample, resulting in a color reaction. The concentration is determined by allocating the color to a value on a reference scale.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service