Skip to Content
Merck
All Photos(1)

Key Documents

1.14412

Supelco

Zinc Test Kit, colorimetric

0.1-5 mg/L (Zn), MQuant®

Synonym(s):

Zinc Visual Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

120 TESTS
$299.00

$299.00


Estimated to ship on23 May 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
120 TESTS
$299.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$299.00


Estimated to ship on23 May 2025


Request a Bulk OrderRequest more information

Product Name

Zinc Test, colorimetric, 0.1-5 mg/L (Zn), for use with MQUANT®

specific analyte(s)

zinc

Quality Level

measuring range

0.1-5 mg/L (Zn)

compatibility

for use with MQUANT®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and comparator block with short test tubes 0.1 - 0.2 - 0.3 - 0.4 - 0.5 - 0.7 - 1 - 2 - 5 mg/l Zn MQuant®
The MQuant® Zinc test (formally MColortest®) measures zinc ions in:
  • Groundwater and surface water
  • Drinking water and mineral water
  • Beverages
  • Industrial water
  • Boiler water
  • Electroplating baths and bath solutions, electroplating wastewater
  • Wastewater and percolating water
This test is not suited for seawater.

Method
In sulfuric solution zinc ions react with thiocyanate ions and brilliant green to form a blue-green complex. The zinc concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card. The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Using the refill pack 1.14782 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system. 

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service