Skip to Content
Merck
All Photos(1)

Key Documents

1.12979

Supelco

Total dietary fiber Kit

sufficient for 100 determinations

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$948.00

$948.00


Estimated to ship on01 June 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
100 TESTS
$948.00

About This Item

UNSPSC Code:
41116118
NACRES:
NA.21

$948.00


Estimated to ship on01 June 2025


Request a Bulk OrderRequest more information

usage

sufficient for 100 determinations

Quality Level

technique(s)

photometry: suitable

storage temp.

2-8°C

General description

reagent kit for 100 determinations

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service