Skip to Content
Merck
All Photos(1)

Key Documents

1.12384

Sigma-Aldrich

Titanium hydride

fine powder 98+

Synonym(s):

Titanium hydride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$182.00
1 KG
$1,330.00

$182.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$182.00
1 KG
$1,330.00

About This Item

Empirical Formula (Hill Notation):
H2Ti
CAS Number:
Molecular Weight:
49.88
MDL number:
UNSPSC Code:
12352306
EC Index Number:
231-726-8
NACRES:
NA.22
grade:
for analytical purposes

$182.00


Estimated to ship on26 May 2025


Request a Bulk Order

grade

for analytical purposes

Quality Level

reaction suitability

reagent type: reductant

mp

>400 °C

transition temp

flash point >400 °C

density

3.76 g/cm3 at 20 °C

bulk density

1400 kg/m3

storage temp.

no temp limit

SMILES string

[Ti+2].[H-].[H-]

InChI

1S/Ti.2H/q+2;2*-1

InChI key

KAZWGWWZKAHTKC-UHFFFAOYSA-N

Analysis Note

Identity (elemental analysis): passes test
Assay (potentiometric): ≥ 98.0 %
H (Hydrogen): 3.6 - 4.4 %

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service