Skip to Content
Merck
All Photos(1)

Key Documents

1.12233

Millipore

L-Lysine monohydrate

for biochemistry

Synonym(s):

L-Lysine monohydrate, Lys

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$154.00

$154.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
$154.00

About This Item

Linear Formula:
NH2(CH2)4CH(NH2)COOH
CAS Number:
Molecular Weight:
146.19 (anhydrous basis)
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-294-2
NACRES:
NA.21

$154.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

form

solid

pH

9-10.5 (20 °C, 80 g/L in H2O)

mp

207-209 °C (decomposes)

solubility

>1500 g/L

bulk density

366 kg/m3

storage temp.

2-8°C

SMILES string

[N+H3][C@@H](CCCCN)C(=O)[O-].O

InChI

1S/C6H14N2O2.H2O/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H2/t5-;/m0./s1

InChI key

HZRUTVAFDWTKGD-JEDNCBNOSA-N

Analysis Note

Assay (Perchloric acid titration, calc. on anhydrous substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance: white, crystalline powder or colorless crystals with characteristic odor
Appearance of solution (100 g/l, water): clear and colorless
Spec. rotation(α 20/D, 50 g/l, hydrochloric acid 1 mol/l, calc. on dried substance): +25.5 to +26.5
Heavy metals (as Pb): ≤ 0.001 %
NH₄ (Ammonium): ≤ 0.01 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.1 %
Water (according to Karl Fischer): 10.0 - 12.0 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service