Skip to Content
Merck
All Photos(1)

Key Documents

1.10329

Millipore

Malachite-green broth (base)

for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74
Pricing and availability is not currently available. Contact Technical Service

Quality Level

form

powder (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.1-7.5 (25 °C in H2O)

application(s)

diagnostic assay manufacturing
food and beverages
hematology
histology
microbiology

storage temp.

15-25°C

suitability

Pseudomonas spp.

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish brown
pH-value (25 °C): 7.1 - 7.5
Inoculum (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): about 1 %
Inoculum (Enterobacter aerogenes ATCC 13048): about 99 %
Enrichment from a mixed inoculum after 20 hours (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): ≥ 50 %
Enrichment from a mixed inoculum after 20 hours (Enterobacter aerogenes ATCC 13048): ≤ 50 %
Enrichment from a mixed inoculum after 44 hours (Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)): ≥ 80 %
Enrichment from a mixed inoculum after 44 hours (Enterobacter aerogenes ATCC 13048): ≤ 20 %
detection with BROLACIN agar

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service