1.10283
GranuCult® plus LB agar (MILLER)
Synonym(s):
LB Agar, LB Agar, Miller Modification, Luria Bertani Agar, Miller Luria Bertani Agar
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
granular
packaging
pkg of 500 g
manufacturer/tradename
GranuCult® plus
storage condition
dry media (Keep tightly closed)
technique(s)
microbiological culture: suitable
color
light yellow to brown
pH
6.8-7.2 (25 °C in H2O, after autoclaving)
application(s)
microbiology
pharmaceutical
storage temp.
15-25°C
suitability
Escherichia coli
Related Categories
General description
Application
Features and Benefits
- GranuCult® offers superior granulated culture media
- Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment
- Excellent wettability, solubility, and free-flowing properties
- Convenient, with minimal component separation and clumping, even under warm or humid conditions
- High batch-to-batch reproducibility
- Prolonged shelf life of up to five years
- High number of test strains exceeding all regulatory demands
- Granulation technology allows many supplements to be included, with no need to add these separately
Linkage
Analysis Note
Appearance (color): yellowish-brown
pH-value (25 °C): 6.8 - 7.2
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good
Growth (Escherichia coli ATCC 11775 (WDCM 00090)): good
Growth (Escherichia coli ATCC 53868 (DH5a)): good
Growth (Escherichia coli ATCC 39403 (JM103)): good
Growth (Escherichia coli DSM 6897 (DH5alpha)): good
Growth (Escherichia coli ATCC 47092 (BL21)): good
Incubation: 24 hours at 35 °C, aerobic
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service