Skip to Content
Merck
All Photos(1)

Key Documents

110116

Sigma-Aldrich

β1-2,3,4,6-N-Acetylglucosaminidase, Streptococcus pneumoniae, Recombinant, E. coli

Synonym(s):

β1-2,3,4,6-N-Acetylglucosaminidase, Streptococcus pneumoniae, Recombinant, E. coli, N-Acetyl-β-D-glycosaminide-N-acetylglucosaminohydrolase

Sign Into View Organizational & Contract Pricing

Select a Size

1 U
$753.00

$753.00


Estimated to ship on31 May 2025



Select a Size

Change View
1 U
$753.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.42

$753.00


Estimated to ship on31 May 2025


recombinant

expressed in E. coli

Quality Level

form

liquid

specific activity

≥35 units/mL
≥80 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

Endo-α-N-acetylgalactosaminidase, α- and β-galactosidase, neuraminidases, proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service