Skip to Content
Merck
All Photos(1)

Key Documents

1.09953

Supelco

Zinc standard

1000 mg Zn, (ZnCl₂ in 0.06% HCl) Titrisol®

Synonym(s):

Zinc Reference Standard for ICP/AAS

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
$67.90

$67.90


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1 AMP
$67.90

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$67.90


Available to ship on31 March 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

product line

Titrisol®

form

concentrate

concentration

1000 mg Zn in 0.06% HCl

technique(s)

AAS: suitable
ICP: suitable

pH

2.0 (20 °C in H2O)

density

1.04 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Application

Zinc standard can be used as a stock standard while determining concentration of zinc in disordered bone serum samples by flame atomic absorption spectroscopy. [1]

Features and Benefits

  • Titrisol® offers a wide range of concentrated single-element standards.
  • Dilute the concentrated solution to achieve the desired standard concentration.
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST).
  • Supplied in easy-to-handle plastic ampoules.
  • Accompanied with a batch-specific certificate of analysis (COA) containing information on the NIST batch and minimum shelf life.
  • Provides reliable and reproducible results.

Analysis Note

content (Zn): 1000 mg / Ampoule
Determination method: Complexometric titration.
(traceable of NIST - SRM 682)
Accuracy of the method: +/- 2 mg

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Brugger et al.
The British journal of nutrition, 121(8), 849-858 (2019-01-30)
The effects of subclinical Zn deficiency on depletion and redistribution of body Zn were studied in weaned piglets. Forty-eight weaned piglets (German-Large-White×Land-Race×Piétrain; 50 % female, 50 % male-castrated; body weight 8·5 (sd 0·27) kg) were fed restrictively (450 g/d) a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service