Skip to Content
Merck
All Photos(1)

Key Documents

1.09926

Supelco

Molybdenum standard

1000 mg Mo, [(NH₄)₆Mo₇O₂₄ 4H₂O in 0.7% NH₄OH] Titrisol®

Sign Into View Organizational & Contract Pricing

Select a Size

1 AMP
$67.90

$67.90


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
1 AMP
$67.90

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$67.90


Estimated to ship on20 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

Titrisol®

form

concentrate

concentration

1000 mg Mo

technique(s)

AAS: suitable
ICP: suitable

pH

9.2 (20 °C in H2O)

density

1.02 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Application

Molybdenumstandard is used as a standard solution to determine the concentration ofmolybdenum in silicate geological samples using Flame Atomic Absorptionspectrometry. [1]

Features and Benefits

  • Titrisol® offers a wide range of concentrated single-element standards.
  • Dilute the concentrated solution to achieve the desired standard concentration.
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST).
  • Supplied in easy-to-handle plastic ampoules.
  • Accompanied with a batch-specific certificate of analysis (COA) containing information on the NIST batch and minimum shelf life.
  • Provides reliable and reproducible results.

Analysis Note

content (Mo): 1000 mg / Ampoule
Determination method:Titration with lead(II)nitrate solution .
(traceable to NIST - SRM 682)
Accuracy of the method: +/- 5 mg

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service