Skip to Content
Merck
All Photos(1)

Key Documents

1.09925

Supelco

Potassium bromate solution

c(KBrO₃) = 1/₆₀ mol/l (0.1 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302

Product Name

Potassium bromate solution, for 1000 ml, c(KBrO₃) = 1/₆₀ mol/l (0.1 N) Titrisol®

Quality Level

product line

Titrisol®

form

liquid

pH

5.6 (20 °C in H2O)

density

1.04 g/cm3 at 20 °C

storage temp.

15-25°C

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Ox. Liq. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Photometric assessment of bromate in drinking water samples using Spectroquant® photometer and test kits, along with 3,3'-dimethylnaphthidine and potassium iodide.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service