Skip to Content
Merck
All Photos(1)

Key Documents

1.09799

Supelco

p-Xylene

reference substance for gas chromatography

Synonym(s):

p-Xylene, 1,4-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(CH3)2
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-396-5

vapor pressure

8.7 hPa ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

540 °C

potency

3910 mg/kg LD50, oral (Rat)

expl. lim.

1.1-7 % (v/v)

technique(s)

gas chromatography (GC): suitable

bp

138.4 °C/1013 hPa

mp

13.3 °C

transition temp

flash point 24 °C

solubility

0.2 g/L

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3

InChI key

URLKBWYHVLBVBO-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 99.5
Identity (IR-spectrum): conforms

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service