Skip to Content
Merck
All Photos(3)

Documents

1.09629

Supelco

tert-Butanol

for analysis EMSURE® ACS

Synonym(s):

tert-Butanol, tert-Butyl alcohol, 2-Methyl-2-propanol, Trimethylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COH
CAS Number:
Molecular Weight:
74.12
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-889-7
NACRES:
NB.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

40.7 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.5% (GC)

form

solid

autoignition temp.

490 °C

expl. lim.

2.3-8.0 % (v/v)

impurities

≤0.00005% Al (Aluminium)
≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤0.01% Aldehydes and ketones (as formaldehyde)
≤0.1% Water

evapn. residue

≤0.001%

color

clearAPHA: ≤10

pH

7 (20 °C)

bp

81-83 °C/1013 hPa

mp

24-25 °C

transition temp

flash point 14 °C

density

0.78 g/cm3 at 20 °C

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

storage temp.

2-30°C

InChI

1S/C4H10O/c1-4(2,3)5/h5H,1-3H3

InChI key

DKGAVHZHDRPRBM-UHFFFAOYSA-N

Related Categories

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application


  • Elucidating and Tuning Catalytic Sites on Zirconium- and Aluminum-Containing Nodes: Investigates the application of tert-butanol in the tuning of catalytic sites within metal-organic frameworks, essential for advancements in material science and catalysis (Yang & Gates, 2021).

  • Photocatalytic mechanism and performance of a novel wool flake-BiFeO3nanosheet-TiO2 core-shell-structured composite photocatalyst: Describes how tert-butanol acts as a solvent in the synthesis of complex photocatalysts, enhancing their efficiency in environmental cleanup applications (Chen et al., 2021).

  • Effect of cross-linked enzyme aggregate strategy on characterization of sn-1,3 extracellular lipase: Demonstrates the role of tert-butanol in enzyme immobilization techniques to improve the stability and activity of lipases, vital for biotechnological applications (Zhu et al., 2021).

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Acidity: ≤ 0.0005 meq/g
Alkalinity: ≤ 0.0002 meq/g
Boiling range (81-83°C): ≥ 95 % (v/v)
Aldehydes and ketones (as formaldehyde): ≤ 0.01 %
Melting point: 24.0 - 26.0 °C
Al (Aluminium): ≤ 0.00005 %
B (Boron): ≤ 0.000002 %
Ba (Barium): ≤ 0.00001 %
Ca (Calcium): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.000005 %
Co (Cobalt): ≤ 0.000002 %
Cr (Chromium): ≤ 0.000002 %
Cu (Copper): ≤ 0.000002 %
Fe (Iron): ≤ 0.00001 %
Mg (Magnesium): ≤ 0.00001 %
Mn (Manganese): ≤ 0.000002 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.00001 %
Sn (Tin): ≤ 0.00001 %
Zn (Zinc): ≤ 0.00001 %
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.1 %
Conforms to ACS, Ph Eur-reagent

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service