1.09628
GranuCult® Buffered Listeria Enrichment Broth (base)
pkg of 500 g, FDA (BAM), for Listeria spp.
Synonym(s):
Listeria Enrichment Broth, Listeria Selective Broth
About This Item
Recommended Products
reg. compliance
FDA (BAM)
Quality Level
form
medium granules (dehydrated (DCM))
packaging
pkg of 500 g
manufacturer/tradename
GranuCult®
technique(s)
microbiological culture: suitable
pH
7.1-7.5 (25 °C in H2O)
application(s)
food and beverages
microbiology
pathogen testing
storage temp.
15-25°C
suitability
Listeria spp. (spp.)
General description
Application
Linkage
Analysis Note
Appearance (colour): yellowish-brown
pH-value (25 °C): 7.1 - 7.5
Inoculum on reference medium (Listeria monocytogenes ATCC 19114 (WDCM 00110)):
Inoculum on reference medium (Listeria monocytogenes ATCC 19111 (WDCM 00020)):
Inoculum on reference medium (Listeria innocua ATCC 33090 (WDCM 00017)):
Inoculum on reference medium (Escherichia coli ATCC 25922 (WDCM 00013)):
Inoculum on reference medium (Enterococcus faecalis ATCC 19433 (WDCM 00009)):
Colony count (Listeria monocytogenes ATCC 19114 (WDCM 00110)): ≥ 1E+08 CFU/ml
Colony count (Listeria monocytogenes ATCC 19111 (WDCM 00020)): ≥ 1E+08 CFU/ml
Colony count (Listeria innocua ATCC 33090 (WDCM 00017)): ≥ 1E+08 CFU/ml
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): no growth
Growth (Enterococcus faecalis ATCC 19433 (WDCM 00009)): passes test
Incubation 48 h at 30 °C
Legal Information
Disclaimer
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service