Skip to Content
Merck
All Photos(1)

Key Documents

1.09479

Supelco

Multi-element standard III dissolved in oil

900 ppm: Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn Certipur®

Synonym(s):

Oil-dissolved Multi-element Standard for ICP/AAS

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$1,600.00

$1,600.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$1,600.00

About This Item

UNSPSC Code:
41116107
EC Index Number:
232-384-2
NACRES:
NA.24

$1,600.00


Estimated to ship on29 May 2025


Request a Bulk Order

grade

certified reference material

Quality Level

product line

Certipur®

concentration

900 ppm in oil

technique(s)

AAS: suitable
ICP: suitable

kinematic viscosity

200 cSt(40 °C)

bp

<300 °C

density

1.1 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

Application

Multi-element standard III dissolved in oil is intended to use as a calibration standard for elemental analysis through ICP and AAS methods.

Analysis Note

Ag (Silver): 885 – 915
Al (Aluminium): 885 – 915
B (Boron): 885 – 915
Ba (Barium): 885 – 915
Ca (Calcium): 885 – 915
Cd (Cadmium): 885 – 915
Cr (Chromium): 885 – 915
Cu (Copper): 885 – 915
Fe (Iron): 885 – 915
Mg (Magnesium): 885 – 915
Mo (Molybdenum): 885 – 915
Mn (Manganese): 885 – 915
Na (Sodium): 885 – 915
Ni (Nickel): 885 – 915
P (phosphorus): 885 – 915
Pb (Lead): 885 – 915
Si (Silicon): 885 – 915
Sn (Tin): 885 – 915
Ti (Titanium): 885 – 915
V (Vanadium): 885 – 915
Zn (Zinc): 885 – 915
Determination methods: ICP OES, ICP MS, GFAA

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service