Skip to Content
Merck
All Photos(1)

Key Documents

1.09237

Millipore

Sampling Head

for use with MAS-100 CG Ex®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403
NACRES:
NA.74
Pricing and availability is not currently available. Contact Technical Service

material

anodized aluminum

Agency

non-compendial

description

pressure head

feature

no. of holes 300 autoclavable

manufacturer/tradename

MAS-100 CG Ex®

parameter

20 bar max. pressure (certified for use as a pressure vessel)

application(s)

compressed gas monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100 CG Ex®

storage temp.

no temp limit

General description

Sampling Head SHLF-CG is an additional accessory designed to be specifically used for MAS-100 CG Ex®. The MAS-100 CG Ex® has been specially developed for monitoring and measuring the microbiological contamination of compressed gases. The sampling method is based on the tried-and-tested Andersen impaction principle and accommodates the use of standard 90 to 100 mm Petri dishes. The included sieve plate is identical with other perforated lids for air samplers, such as MAS-100 NT® providing 300 holes of a 0.6 mm diameter resulting in a D50 of 1.1 μm at a flow rate of 100 liters/min. The sampling head is certified for use as pressure vessel up to a maximum pressure of 10 bar. The sampling head is made from anodized aluminum with tri-clamps made of stainless steel and can be sterilized. The instrument is an apparatus of category 3 according to EN 600079-14:1997. It can only be used in Zone 2 for gas groups IIA, IIB, IIC, compromising flammable substances which pose an explosion hazard in the temperature classes T1 to T6.

Application

The microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as for Food, Beverage and Environmental applications.

Legal Information

MAS-100 CG Ex is a registered trademark of MBV AG
MAS-100 NT is a registered trademark of MBV AG

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service