Skip to Content
Merck
All Photos(2)

Key Documents

1.09211

Sigma-Aldrich

Azure II (C.I.52010/52015)

for microscopy

Synonym(s):

Azure II, 3,7 Bis(dimethylamino)phenothiazin-5-ium chloride and 3-(Dimethylamino)-7-(methylamino)-phenothiazin-5-ium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$68.10

$68.10


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 G
$68.10

About This Item

Linear Formula:
C16H18N3S · C15H16N3S ·2 Cl
CAS Number:
Molecular Weight:
625.68
Colour Index Number:
52010/52015
MDL number:
UNSPSC Code:
41116130
NACRES:
NA.21

$68.10


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

form

solid

IVD

for in vitro diagnostic use

pH

2.9 (20 °C, 10 g/L in H2O)

solubility

100 g/L

bulk density

610 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

SMILES string

[s+]1c2c(nc3c1cc(cc3)N(CC)CC)ccc(c2)N(CC)CC
[Cl-].[Cl-].CNc1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](\C)C.CN(C)c4ccc5N=C6C=C\C(C=C6Sc5c4)=[N+](\C)C

InChI

1S/C16H18N3S.C15H15N3S.2ClH/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;1-16-10-4-6-12-14(8-10)19-15-9-11(18(2)3)5-7-13(15)17-12;;/h5-10H,1-4H3;4-9H,1-3H3;2*1H/q+1;;;/p-1

InChI key

YOWZJZJLXUQHGF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Azure II (C.I. 52010/ 52015), is a dry dye for the preparation of a color staining solution used for the staining of blood and bone marrow smears of human origin. The present dye Azure II (C.I.52010/52015) - for microscopy is a defined dye mixture of azure I and methylene blue (1+1), for use in hematological stainings by addition of eosin Y. The staining of the nuclei is due of the molecular interaction of the Eosin Y dye and a complex of Azur B with DNA. Both dyes assemble to an Eosin Y - Azur B-DNA complex and the intensity of the resulting stain depends on the content of Azur B and the ratio of Azur B : Eosin Y.
Furthermore, the resulting stain can vary depending on the influence of fixation, staining times, pH-value of the solutions or buffer substances. The dye is a CE certified IVD product and has a constant dye content and a reliable high quality, which is one of the major precondition for a reproducible diagnostic staining. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service