Skip to Content
Merck
All Photos(1)

Key Documents

1.09115

Supelco

Potassium hydroxide solution

c(KCl) = 0.1 mol/L (1 N) in ethanol, reag. USP, ready-to-use volumetric solution for titration in non-aqueous media, Titripur®

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$122.00
1 L
$128.00
2.5 L
$214.00

$122.00


Available to ship on01 April 2025Details


Request more information

Select a Size

Change View
1 L
$122.00
1 L
$128.00
2.5 L
$214.00

About This Item

UNSPSC Code:
12352305
NACRES:
NA.21

$122.00


Available to ship on01 April 2025Details


Request more information

Product Name

Potassium hydroxide solution in ethanol, c(KOH) = 0.1 mol/l (0.1 N), Titripur®, reag. USP

Agency

reag. USP

Quality Level

product line

Titripur®

form

liquid

autoignition temp.

425 °C (ethanol)

quality

Analyzed in our ISO 17025 accredited QC lab

expl. lim.

3.5-15.0 % (v/v) ethanol)

reaction suitability

reaction type: Acid-base reactions

concentration

0.1 M

technique(s)

titration: suitable

pH

14 (20 °C in H2O)

transition temp

flash point 12 °C (ethanol)

density

0.836 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Voltammetric Determination: Potassium hydroxide solution in ethanol is used in voltammetric determination of antioxidants in various matrices. A recent study demonstrated its application in the voltammetric determination of the antioxidant 2,6-di-tert-butyl-p-cresol (T501) in transformer oil using a graphite electrode. This method enhances the sensitivity and accuracy of antioxidant detection, crucial for ensuring the stability and performance of oils and other industrial fluids (Zhou et al., Analytical Letters, 2021).

Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Analysis Note

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005 mol/L
Measurement uncertainty ± 0.0003 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 84 potassium hydrogen phthalate) by means of volumetric standard potassium hydrogen phthalate (article number 1.02400), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - (ethanol)

Flash Point(C)

13 °C - (ethanol)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service