Skip to Content
Merck

108704

SAFC

1-Thioglycerol

EMPROVE® ESSENTIAL, NF

Pharma Manufacturing

Synonym(s):

α-Monothioglycerol, α-Thioglycerol, 3-Mercapto-1,2-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
108.16
Beilstein:
1732046
EC Number:
MDL number:
UNSPSC Code:
12352105
Pricing and availability is not currently available. Contact Technical Service

biological source

synthetic

Quality Level

Agency

NF

form

liquid

packaging

pkg of 500g (1087040500)
pkg of 50g (1087040050)

refractive index

n20/D 1.527 (lit.)

bp

118 °C/5 mmHg (lit.)

density

1.25 g/mL at 25 °C (lit.)

suitability

suitable for manufacturing use

SMILES string

OCC(O)CS

InChI

1S/C3H8O2S/c4-1-3(5)2-6/h3-6H,1-2H2

InChI key

PJUIMOJAAPLTRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service