Skip to Content
Merck
All Photos(1)

Documents

1.08409

Supelco

Ethylendiamintetraacetic acid magnesium dipotassium salt

for analysis

Synonym(s):

Ethylendiamintetraacetic acid magnesium dipotassium salt, Ethylenediaminetetraacetic acid dipotassium magnesium salt, Ethylenedinitrilotetraacetic acid dipotassium magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12K2MgN2O8 · 2H2O
Molecular Weight:
426.74
UNSPSC Code:
12352107

Quality Level

form

solid

reaction suitability

reaction type: Complexometric reactions

technique(s)

titration: suitable

pH

6.0-9.0 (20 °C, 50 g/L in H2O)

solubility

200 g/L

bulk density

730 kg/m3

storage temp.

no temp limit

Analysis Note

Assay (complexometric, calc. on anhydrous substance): ≥ 98 %
Identity (IR-spectrum): passes test
Heavy metals (as Pb): ≤ 0.0005 %
Fe (Iron): ≤ 0.001 %
Free Magnesium: ≤ 0.01 %
Free EDTA: ≤ 0.05 %
Water (according to Karl Fischer): 7.5 - 9.5 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service