Skip to Content
Merck
All Photos(1)

Key Documents

1.08320

Millipore

N-[Tris(hydroxymethyl)methyl]-3-aminopropanesulfonic acid

pKa (20 °C)8.40, ≥99% (alkalimetric)

Synonym(s):

N-[Tris(hydroxymethyl)methyl]-3-aminopropanesulfonic acid, TAPS

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$374.00

$374.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
$374.00

About This Item

CAS Number:
Molecular Weight:
243.27
MDL number:
UNSPSC Code:
12161703
EC Index Number:
249-954-1
NACRES:
NA.05

$374.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥99% (alkalimetric)

pKa (20 °C)

8.40

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])CCC[N+H2]C(CO)(CO)CO

InChI

1S/C7H17NO6S/c9-4-7(5-10,6-11)8-2-1-3-15(12,13)14/h8-11H,1-6H2,(H,12,13,14)

InChI key

YNLCVAQJIKOXER-UHFFFAOYSA-N

General description

buffer substance TAPS

Application

N-[Tris(hydroxymethyl)methyl]-3-aminopropanesulfonic acid buffer substance TAPS. CAS 29915-38-6, molar mass 243.27 g/mol.

Analysis Note

Assay (alkalimetric)≥ 99 %Identity (IR-spectrum)passes testUV-Absorption (250 nm, 20 g/l, 1 cm, water)≤ 0.05Chloride (Cl)≤ 0.005 %Sulphate (SO₄)≤ 0.005 %Heavy metals (as Pb)≤ 0.0005 %Na (Sodium)≤ 0.005 %Loss on Drying (105°C)≤ 0.2 %Biological buffer, pKa at 20 °C8.40

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service