Skip to Content
Merck
All Photos(1)

Key Documents

1.08159

Supelco

Holmium perchlorate standard solution

for the calibration of spectrophotometers

Synonym(s):

Calibration standard for UV-VIS Spectrophotometer, Holmium standard for UV-VIS Spectrophotometer

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$834.00

$834.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$834.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$834.00


Estimated to ship on27 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

technique(s)

UV/Vis spectroscopy: suitable

density

1.136 g/cm3 at 20 °C

application(s)

environmental
pharmaceutical

format

mixture

storage temp.

15-25°C

Application

  • Holmium perchlorate standard solution for use in calibration of spectrophotometers
  • May be used to calibrate a newly installed instrument or after a major repair of an instrument
  • Applicable for quality control checks and system suitability tests

Analysis Note

The principal absorption bands (2nm slit, 30 nm/min. scan speed) of this solution are at the following wavelengths:
λ max: 241.1 ± 0.4 nm
:
:
:
:
:
:
:
:

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 2 - Skin Corr. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service