Skip to Content
Merck
All Photos(1)

Key Documents

1.07916

Supelco

Spectrosolv PSS

for the ACIDORAPID mineralizer

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$1,580.00

$1,580.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
1 L
$1,580.00

About This Item

UNSPSC Code:
12352402
NACRES:
NA.21

$1,580.00


Estimated to ship on16 April 2025


Request a Bulk Order

Quality Level

form

liquid

pH

(20 °C, (68 °F) strongly acid)

solubility

water: soluble

density

1.78 g/cm3 at 20 °C

cation traces

Al: ≤0.2 ppm
Ba: ≤0.2 ppm
Cd: ≤0.1 ppm
Co: ≤0.1 ppm
Cr: ≤0.1 ppm
Fe: ≤0.1 ppm
K: ≤0.1 ppm
Li: ≤0.1 ppm
Mg: ≤0.1 ppm
Mn: ≤0.1 ppm
Na: ≤0.2 ppm
Ni: ≤0.1 ppm
Pb: ≤0.1 ppm
Sr: ≤0.1 ppm
Tl: ≤0.1 ppm
Zn: ≤0.2 ppm

storage temp.

2-30°C

Analysis Note

Assay (H₃PO₄): 40.9 - 43.7 %
Assay (H₂SO₄): 47.3 - 50.0 %
Al (Aluminium): ≤ 0.2 ppm
Ba (Barium): ≤ 0.2 ppm
Cd (Cadmium): ≤ 0.1 ppm
Co (Cobalt): ≤ 0.1 ppm
Cr (Chromium): ≤ 0.1 ppm
Fe (Iron): ≤ 0.1 ppm
K (Potassium): ≤ 0.1 ppm
Li (Lithium): ≤ 0.1 ppm
Mg (Magnesium): ≤ 0.1 ppm
Mn (Manganese): ≤ 0.1 ppm
Na (Sodium): ≤ 0.2 ppm
Ni (Nickel): ≤ 0.1 ppm
Pb (Lead): ≤ 0.1 ppm
Sr (Strontium): ≤ 0.1 ppm
Tl (Thallium): ≤ 0.1 ppm
Zn (Zinc): ≤ 0.2 ppm

Legal Information

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service