Skip to Content
Merck
All Photos(1)

Key Documents

1.07341

Supelco

Hexachloroplatinic(IV) acid solution

10%, (3.8% Pt) GR for analysis

Synonym(s):

HCl6Pt Solution

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$371.00
25 ML
$1,510.00

$371.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
$371.00
25 ML
$1,510.00

About This Item

UNSPSC Code:
12352301
NACRES:
NA.21

$371.00


Estimated to ship on31 May 2025


Request a Bulk Order

Product Name

Hexachloroplatinic(IV) acid solution about 10%, (3.8% Pt) GR for analysis

Quality Level

Assay

≥10.0% (gravimetric)

form

liquid

concentration

≥5-10% (Hexachloroplatinic(IV) acid)

pH

0.6 (20 °C in H2O)

solubility

water: soluble

density

1.06 g/cm3 at 20 °C

application(s)

general analytical

storage temp.

15-25°C

SMILES string

[H+].[Cl-][Pt+4]([Cl-])([Cl-])([Cl-])([Cl-])[Cl-]

InChI

InChI=1S/6ClH.Pt/h6*1H;/q;;;;;;+4/p-4

InChI key

GBFHNZZOZWQQPA-UHFFFAOYSA-J

Application


  • Aggregated structure analysis of polymer-protected platinum/ruthenium colloidal dispersions using EXAFS, HRTEM, and electron diffraction measurements: This research highlights the application of Hexachloroplatinic(IV) acid solution in the synthesis of advanced platinum/ruthenium catalysts. The study utilizes a 10% solution to prepare polymer-protected colloidal dispersions, instrumental in catalyzing reactions under environmentally benign conditions. The catalysts produced demonstrate improved activity and selectivity, key attributes for applications ranging from pharmaceutical synthesis to material science innovation (Harada M, Toshima N, Yoshida K, Isoda S, 2005).

Analysis Note

Assay (gravimetrically): ≥ 10.0 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service