Skip to Content
Merck
All Photos(1)

Key Documents

1.07243

Supelco

1,2-Phenylenediamine

GR for analysis

Synonym(s):

1,2-Phenylenediamine, 1,2-Diaminobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$162.00

$162.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$162.00

About This Item

Linear Formula:
1,2-(NH2)2C6H4
CAS Number:
Molecular Weight:
108.14
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-430-6
NACRES:
NB.21

$162.00


Check Cart for Availability

Request a Bulk Order

vapor pressure

0.013 hPa ( 20 °C)

Quality Level

form

solid

autoignition temp.

540 °C

expl. lim.

1.5 % (v/v)

pH

7-8 (20 °C, 50 g/L in H2O)

bp

257 °C/1013 hPa

mp

99-103 °C

transition temp

flash point 110 °C

solubility

54 g/L

density

1.14 g/cm3 at 20 °C

bulk density

530 kg/m3

storage temp.

2-30°C

SMILES string

Nc1c(cccc1)N

InChI

1S/C6H8N2/c7-5-3-1-2-4-6(5)8/h1-4H,7-8H2

InChI key

GEYOCULIXLDCMW-UHFFFAOYSA-N

Application



  • Synthesis of imines from the coupling reaction of alcohols and amines catalyzed by phosphine-free cobalt(II) complexes.: This study explores the use of 1,2-phenylenediamine in catalytic systems, demonstrating its efficacy in synthesizing imines, which are pivotal in various synthetic applications (Mahato et al., 2024).


  • Copper(II) Driven Fluorescence switch-on Detection of Ovalbumin and GSH Using a Pyridoxal 5′-phosphate Derived Tetradentate Schiff Base and its Applications.: Research demonstrates the application of 1,2-phenylenediamine-based ligands in creating sensitive fluorescent probes for bioanalytical applications (Dhanshri et al., 2024).


Analysis Note

Assay (HClO₄): ≥ 99.0 %
Identity (IR): passes test
Melting range (lower value): ≥ 99 °C
Melting range (upper value): ≤ 103 °C
Sulfated ash: ≤ 0.25 %
Water (K. F.): ≤ 1.0 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service