Skip to Content
Merck
All Photos(1)

Key Documents

1.07047

Supelco

1,4-Dioxane

for analysis EMPARTA® ACS

Synonym(s):

1,4-Dioxane, Diethylene oxide, Dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102551
MDL number:
UNSPSC Code:
12352005
grade:
ACS reagent
bp:
100-102 °C (lit.)
vapor pressure:
27 mmHg ( 20 °C)
40 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

ACS reagent

vapor density

3 (vs air)

vapor pressure

27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

autoignition temp.

356 °F

expl. lim.

22 %

dilution

(for routine analytical applications)

refractive index

n20/D 1.422 (lit.)

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

storage temp.

20-25°C

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service